Assay Details
Target Gene Details
Entrez Gene ID: | 72322 |
Gene Name: | exportin 5 |
Gene Aliases: |
2410004H11Rik, 2700038C24Rik, AI648907, AW549301, Exp5, RanBp21, mKIAA1291 |
Location: |
Chr.17:46202855-46242299 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 15 - Intron 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Xpo5 | NM_028198.2 | NP_082474.1 | ||
| AF343581.1 | AAK68050.1 | |||
| BC131660.1 | ||||
| BC131661.1 |
More Information
Set Membership: |
|

Genomic Map