Assay Details
Target Gene Details
Entrez Gene ID: | 93687 |
Gene Name: | casein kinase 1, alpha 1 |
Gene Aliases: |
2610208K14Rik, 4632404G05Rik, 5430427P18Rik, CK1a, Csnk1a |
Location: |
Chr.18:61555165-61590315 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Csnk1a1 | NM_146087.2 | 9 | 1078 | NP_666199.1 |
| AK076070.1 | 9 | 1345 | BAC36161.1 | |
| AK076264.1 | 9 | 1337 | ||
| AK078398.1 | 9 | 1327 | BAC37255.1 | |
| AK141421.1 | 9 | 1333 | ||
| AK146873.1 | 9 | 1348 | BAE27496.1 | |
| AK147165.1 | 9 | 1323 | BAE27730.1 | |
| AK152387.1 | 9 | 1323 | BAE31175.1 | |
| AK152793.1 | 9 | 1343 | BAE31499.1 | |
| AK159939.1 | 6 | 547 | BAE35497.1 | |
| AK166318.1 | 9 | 1338 | BAE38701.1 | |
| AK166429.1 | 9 | 1320 | ||
| AK167550.1 | 9 | 1346 | ||
| AK182366.1 | 1 | 66 | ||
| AK191355.1 | 1 | 66 | ||
| AK191813.1 | 1 | 66 | ||
| AK192016.1 | 1 | 66 | ||
| AK194970.1 | 1 | 66 | ||
| AK195332.1 | 1 | 66 | ||
| AK196802.1 | 1 | 66 | ||
| AK201811.1 | 1 | 66 | ||
| AK203785.1 | 1 | 66 | ||
| AK207201.1 | 1 | 66 | ||
| AK212753.1 | 1 | 66 | ||
| AK217167.1 | 1 | 66 | ||
| BC002171.1 | 3 | 226 | AAH02171.1 | |
| BC019740.1 | 9 | 1078 | AAH19740.1 | |
| BC025439.1 | 9 | 1025 | AAH25439.1 | |
| BC048081.1 | 9 | 1111 | AAH48081.1 | |
| X90945.1 | CAA62440.1 |
More Information
Set Membership: |
|

Genomic Map