Assay Details
Target Gene Details
Entrez Gene ID: | 72322 |
Gene Name: | exportin 5 |
Gene Aliases: |
2410004H11Rik, 2700038C24Rik, AI648907, AW549301, Exp5, RanBp21, mKIAA1291 |
Location: |
Chr.17:46202855-46242299 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Xpo5 | NM_028198.2 | 5 | 554 | NP_082474.1 |
| AF343581.1 | 5 | 554 | AAK68050.1 | |
| AK010389.1 | 5 | 585 | BAB26904.1 | |
| AK028747.1 | 1 | 1647 | ||
| BC055455.1 | 5 | 592 | AAH55455.1 | |
| BC131660.1 | 5 | 550 | ||
| BC131661.1 | 5 | 550 |
More Information
Set Membership: |
|

Genomic Map