Assay Details
Target Gene Details
Entrez Gene ID: | 52563 |
Gene Name: | CDC23 cell division cycle 23 |
Gene Aliases: |
6030435O18, D18Ertd243e |
Location: |
Chr.18:34630946-34651745 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdc23 | NM_178347.4 | NP_848124.1 | ||
| AK031459.1 | BAC27415.1 | |||
| AK038523.1 | BAC30026.1 | |||
| AK047789.1 | ||||
| AK048246.1 | BAC33283.1 | |||
| AK052567.1 | ||||
| AK075948.1 | ||||
| AK079681.1 | ||||
| AK081461.1 | BAC38224.1 | |||
| BC059013.1 | AAH59013.1 |
More Information
Set Membership: |
|

Genomic Map