Assay Details
Target Gene Details
Entrez Gene ID: | 225164 |
Gene Name: | mindbomb homolog 1 (Drosophila) |
Gene Aliases: |
DIP-1, E430019M12Rik, Mib, mKIAA1323 |
Location: |
Chr.18:10725625-10818578 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mib1 | NM_144860.2 | 21 | 3633 | NP_659109.2 |
| AK053035.1 | 14 | 1856 | BAC35245.1 | |
| AK080847.1 | BAC38042.1 | |||
| AK081370.1 | 2 | 93 | ||
| AK129331.1 | 4 | 364 | BAC98141.1 | |
| AK161848.1 | 6 | 729 | BAE36604.1 | |
| AY147848.1 | 21 | 3132 | AAN18022.1 | |
| AY149907.1 | 21 | 2953 | AAN75492.1 | |
| AY245539.1 | 21 | 3633 | AAO91933.1 | |
| AY974091.1 | 21 | 2926 | AAX84653.1 | |
| BC011287.1 | 10 | 1230 | AAH11287.1 | |
| BC083072.1 | 11 | 1311 | AAH83072.1 | |
| BC131662.1 | 22 | 3099 |
More Information
Set Membership: |
|

Genomic Map