Assay Details
Target Gene Details
Entrez Gene ID: | 19038 |
Gene Name: | peptidylprolyl isomerase C |
Gene Aliases: |
CyP-20c |
Location: |
Chr.18:53406341-53418007 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppic | NM_008908.4 | NP_032934.1 | ||
| AK131931.1 | BAE20880.1 | |||
| AK150654.1 | BAE29741.1 | |||
| AK151449.1 | BAE30409.1 | |||
| AK168911.1 | BAE40724.1 | |||
| BC025861.1 | AAH25861.1 | |||
| M74227.1 | AAA37511.1 |
More Information
Set Membership: |
|

Genomic Map