Assay Details
Target Gene Details
Entrez Gene ID: | 433181 |
Gene Name: | serine peptidase inhibitor, Kazal type 11 |
Gene Aliases: |
9230112K01Rik |
Location: |
Chr.18:44190045-44196177 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Spink11 | NM_001048217.3 | NP_001041682.2 | ||
| AK136741.1 | BAE23116.1 | |||
| BC147524.1 | ||||
| BC147541.1 | ||||
| DQ437331.1 |
More Information
Set Membership: |
|

Genomic Map