Assay Details
Target Gene Details
Entrez Gene ID: | 100384868 |
Gene Name: | predicted gene, 37013 |
Gene Aliases: |
Pcdha4-g |
Location: |
Chr.18:36952648-37841873 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm37013 | NM_001174154.2 | NP_001167625.1 |
Target Gene Details
Entrez Gene ID: | 103611158 |
Gene Name: | predicted gene, 38666 |
Gene Aliases: |
Pcdha6-g |
Location: |
Chr.18:36967631-37841873 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm38666 | NM_001301256.1 | NP_001288185.1 |
Target Gene Details
Entrez Gene ID: | 103611159 |
Gene Name: | protocadherin alpha 7-gamma |
Gene Aliases: |
- |
Location: |
Chr.18:36973802-37841873 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pcdha7-g | NM_001301259.1 | NP_001288188.1 |
Target Gene Details
Entrez Gene ID: | 24074 |
Gene Name: | TAF7 RNA polymerase II, TATA box binding protein (TBP)-associated factor |
Gene Aliases: |
55kDa, AI607308, BB007175, TAFII55, Taf2f |
Location: |
Chr.18:37640491-37644204 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Taf7 | NM_175770.4 | NP_786964.1 | ||
| AF144562.1 | AAD46767.1 | |||
| AK029380.1 | BAC26429.1 | |||
| AK075700.1 | BAC35898.1 | |||
| AK076362.1 | BAC36313.1 | |||
| AK089054.1 | BAC40723.2 | |||
| BC029673.1 | AAH29673.1 | |||
| DV656353.1 |
More Information
Set Membership: |
|

Genomic Map