Assay Details
Target Gene Details
Entrez Gene ID: | 56736 |
Gene Name: | ring finger protein 14 |
Gene Aliases: |
2310075C09Rik, 2610005D23Rik, AA986456, AU041447, D18Ertd188e, D7Bwg0165e, Triad2 |
Location: |
Chr.18:38296635-38317852 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rnf14 | NM_001164621.1 | NP_001158093.1 | ||
| NM_001164622.1 | NP_001158094.1 | |||
| NM_020012.2 | NP_064396.2 | |||
| AF249667.1 | AAF74266.1 | |||
| AF249668.1 | AAF74267.1 | |||
| AK011316.2 | BAB27541.1 | |||
| AK038986.1 | ||||
| BC054841.1 | AAH54841.1 | |||
| BC094250.1 | AAH94250.1 |
More Information
Set Membership: |
|

Genomic Map