Assay Details
Target Gene Details
Entrez Gene ID: | 225280 |
Gene Name: | INO80 complex subunit C |
Gene Aliases: |
D030070L09Rik |
Location: |
Chr.18:24104761-24121819 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ino80c | NM_172625.2 | NP_766213.1 | ||
| AK077290.1 | BAC36733.1 | |||
| AK083718.1 | BAC39004.1 | |||
| BC060961.1 | AAH60961.1 |

Genomic Map