Assay Details
Target Gene Details
Entrez Gene ID: | 73473 |
Gene Name: | IWS1 homolog (S. cerevisiae) |
Gene Aliases: |
1700069O15Rik, 3000008H23, AL117927, AW214353 |
Location: |
Chr.18:32067325-32104331 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 11 - Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Iws1 | NM_173441.3 | NP_775617.1 | ||
| AK006925.1 | BAB24793.1 | |||
| AK028247.1 | BAC25838.1 | |||
| AK033013.1 | ||||
| AK136492.1 | BAE23006.1 | |||
| BC068184.1 | AAH68184.1 |

Genomic Map