Assay Details
Target Gene Details
Entrez Gene ID: | 225283 |
Gene Name: | regulation of nuclear pre-mRNA domain containing 1A |
Gene Aliases: |
C77387, mKIAA4077 |
Location: |
Chr.18:24484958-24530222 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rprd1a | NM_144861.2 | NP_659110.1 | ||
| AK137068.1 | 4 | 545 | ||
| AK161533.1 | 4 | 509 | ||
| AK220475.1 | BAD90282.1 | |||
| BC021395.1 | AAH21395.1 | |||
| BC023084.1 | AAH23084.1 |
More Information
Set Membership: |
|

Genomic Map