Assay Details
Target Gene Details
Entrez Gene ID: | 18596 |
Gene Name: | platelet derived growth factor receptor, beta polypeptide |
Gene Aliases: |
AI528809, CD140b, PDGFR-1, Pdgfr |
Location: |
Chr.18:61045150-61085068 on Build GRCm38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pdgfrb | NM_001146268.1 | 22 | 3525 | NP_001139740.1 |
| NM_008809.2 | 22 | 3522 | NP_032835.2 | |
| AK134785.1 | 23 | 3547 | BAE22283.1 | |
| AK143247.1 | 22 | 3504 | ||
| AK158145.1 | BAE34383.1 | |||
| BC055311.1 | 22 | 3537 | AAH55311.1 | |
| DQ979390.1 | 16 | 2178 | ||
| X04367.1 | 22 | 3231 | CAA27882.1 |
More Information
Set Membership: |
|

Genomic Map