Assay Details
Target Gene Details
Entrez Gene ID: | 71983 |
Gene Name: | transmembrane and coiled-coil domains 6 |
Gene Aliases: |
2410015B03Rik |
Location: |
Chr.18:36735018-36742400 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmco6 | NM_028036.3 | NP_082312.3 | ||
| AK010487.1 | BAB26978.1 | |||
| BC005637.1 | ||||
| BF468256.2 |

Genomic Map