Assay Details
Target Gene Details
Entrez Gene ID: | 225326 |
Gene Name: | phosphoinositide-3-kinase, class 3 |
Gene Aliases: |
5330434F23Rik, Vps34 |
Location: |
Chr.18:30272094-30348126 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pik3c3 | NM_181414.5 | 11 | 1295 | NP_852079.2 |
| AK030578.1 | 11 | 1325 | ||
| AK044997.1 | 11 | 1295 | ||
| AK219216.1 | 2 | 94 | ||
| BC024675.1 | 11 | 1330 | AAH24675.1 | |
| BC057678.1 | 11 | 1298 | AAH57678.1 |
More Information
Set Membership: |
|

Genomic Map