Assay Details
Target Gene Details
Entrez Gene ID: | 71983 |
Gene Name: | transmembrane and coiled-coil domains 6 |
Gene Aliases: |
2410015B03Rik |
Location: |
Chr.18:36735018-36742400 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmco6 | NM_028036.3 | 9 | 1112 | NP_082312.3 |
| AK010487.1 | 9 | 1037 | BAB26978.1 | |
| AK046207.1 | 9 | 1150 | BAC32636.1 | |
| BC005637.1 | 9 | 1114 | ||
| BF468256.2 |

Genomic Map