Assay Details
Target Gene Details
Entrez Gene ID: | 16769 |
Gene Name: | desmoglein 4 |
Gene Aliases: |
CDHF13, lah |
Location: |
Chr.18:20436175-20471821 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dsg4 | NM_181564.2 | 10 | 1480 | NP_853543.1 |
| AB240192.1 | 9 | 1663 | ||
| AY170623.1 | 3 | 428 | AAO41856.1 | |
| AY191584.1 | 10 | 1480 | AAO91793.1 | |
| AY227349.1 | 10 | 1346 | AAP44999.1 |
More Information
Set Membership: |
|

Genomic Map