Assay Details
Target Gene Details
Entrez Gene ID: | 58916 |
Gene Name: | myotilin |
Gene Aliases: |
5530402I04Rik, Ttid |
Location: |
Chr.18:44334053-44355740 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Myot | NM_001033621.3 | NP_001028793.1 | ||
| AF230979.2 | AAF76465.1 | |||
| AK017447.1 | BAB30747.1 | |||
| BC016214.1 | AAH16214.1 | |||
| BC064052.1 | AAH64052.1 |
More Information
Set Membership: |
|

Genomic Map