Assay Details
Target Gene Details
Entrez Gene ID: | 72747 |
Gene Name: | tetratricopeptide repeat domain 39C |
Gene Aliases: |
1700008N02Rik, 2810439F02Rik |
Location: |
Chr.18:12643533-12737052 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ttc39c | NM_028341.4 | 1 | 439 | NP_082617.2 |
| AK013267.1 | 1 | 174 | BAB28758.1 | |
| AK080904.1 | 1 | 628 | BAC38071.1 | |
| BC020021.1 | 1 | 189 | AAH20021.1 |

Genomic Map