Assay Details
Target Gene Details
Entrez Gene ID: | 240216 |
Gene Name: | Riken cDNA E230025N22 gene |
Gene Aliases: |
EG240216 |
Location: |
Chr.18:36683078-36696091 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| E230025N22Rik | NM_172831.2 | NP_766419.2 | ||
| AK087618.1 | BAC39945.1 |

Genomic Map