Assay Details
Target Gene Details
Entrez Gene ID: | 14164 |
Gene Name: | fibroblast growth factor 1 |
Gene Aliases: |
Dffrx, Fam, Fgf-1, Fgfa |
Location: |
Chr.18:38838672-38929470 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fgf1 | NM_010197.3 | NP_034327.1 | ||
| AK035330.1 | BAC29035.1 | |||
| AK142163.1 | ||||
| U67610.1 | AAC52907.1 |
Target Gene Details
Entrez Gene ID: | 545253 |
Gene Name: | predicted gene 5820 |
Gene Aliases: |
EG545253 |
Location: |
Chr.18:38892456-38971274 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm5820 | NM_001033789.2 | 3 | 233 | NP_001028961.1 |
| AK145364.1 | 3 | 234 | BAE26390.1 | |
| BC147564.1 | 3 | 184 |
More Information
Set Membership: |
|

Genomic Map