Assay Details
Target Gene Details
Entrez Gene ID: | 72504 |
Gene Name: | TAF4B RNA polymerase II, TATA box binding protein (TBP)-associated factor |
Gene Aliases: |
105kDa, 2610524B04Rik, 4932409F03Rik, AW987595, TAFII-105, TAFII105, Taf2c2 |
Location: |
Chr.18:14783201-14960424 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Taf4b | NM_001100449.1 | 11 | 2396 | NP_001093919.1 |
More Information
Set Membership: |
|

Genomic Map