Assay Details
Target Gene Details
Entrez Gene ID: | 66556 |
Gene Name: | Dr1 associated protein 1 (negative cofactor 2 alpha) |
Gene Aliases: |
2310074H19Rik, NC2-alpha |
Location: |
Chr.19:5422804-5424979 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Drap1 | NM_001291080.1 | 4 | 532 | NP_001278009.1 |
| NM_024176.2 | 4 | 532 | NP_077138.1 | |
| AK005445.1 | 3 | 267 | ||
| AK010152.1 | 4 | 486 | BAB26737.1 | |
| AK087456.1 | 4 | 358 | BAC39882.1 | |
| AK154251.1 | 3 | 312 | BAE32465.1 | |
| BC002090.1 | 4 | 469 | AAH02090.1 | |
| CT010290.1 | 4 | 267 | CAJ18498.1 |
More Information
Set Membership: |
|

Genomic Map