Assay Details
Target Gene Details
Entrez Gene ID: | 14675 |
Gene Name: | guanine nucleotide binding protein, alpha 14 |
Gene Aliases: |
AU023208 |
Location: |
Chr.19:16435667-16610818 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gna14 | NM_008137.4 | NP_032163.3 | ||
| AK035320.1 | BAC29032.1 | |||
| AK085444.1 | BAC39447.1 | |||
| AK160808.1 | BAE36026.1 | |||
| AK168498.1 | BAE40384.1 | |||
| BC027015.1 | AAH27015.1 | |||
| M80631.1 | AAA83222.1 |
More Information
Set Membership: |
|

Genomic Map