Assay Details
Target Gene Details
Entrez Gene ID: | 225207 |
Gene Name: | zinc finger protein 521 |
Gene Aliases: |
B930086A16Rik, Evi3, Znf521 |
Location: |
Chr.18:13687013-13972787 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp521 | NM_145492.4 | NP_663467.1 | ||
| AK052655.1 | ||||
| AK077697.1 | BAC36964.1 | |||
| AK081096.1 | BAC38132.1 | |||
| AK131130.1 | BAD21380.1 | |||
| AY147406.1 | AAN39839.1 | |||
| BC021376.1 | AAH21376.1 |
More Information
Set Membership: |
|

Genomic Map