Assay Details
Target Gene Details
Entrez Gene ID: | 107239 |
Gene Name: | carnosine synthase 1 |
Gene Aliases: |
AI790298, Atpgd1, mKIAA1394 |
Location: |
Chr.19:4164324-4175479 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Carns1 | NM_134148.2 | 8 | 1403 | NP_598909.2 |
AK129347.1 | 7 | 1560 | BAC98157.1 | |
BC023699.1 | 1 | 98 | AAH23699.1 |
