Assay Details
Target Gene Details
Entrez Gene ID: | 12333 |
Gene Name: | calpain 1 |
Gene Aliases: |
Capa-1, Capa1, mu-calpin |
Location: |
Chr.19:5988545-6015825 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Capn1 | NM_001110504.1 | NP_001103974.1 | ||
| BY034288.1 |
Target Gene Details
Entrez Gene ID: | 100502985 |
Gene Name: | predicted gene 10814 |
Gene Aliases: |
ENSMUSG00000075230 |
Location: |
Chr.19:6012620-6018459 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm10814 | NR_045783.1 | |||
| AK149403.1 | 1 | 604 | BAE28853.1 | |
| AK160921.1 |
More Information
Set Membership: |
|

Genomic Map