Assay Details
Target Gene Details
Entrez Gene ID: | 14764 |
Gene Name: | prostaglandin D2 receptor 2 |
Gene Aliases: |
Crth2, Gpr44, Grp45 |
Location: |
Chr.19:10927861-10942511 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ptgdr2 | NM_009962.3 | NP_034092.1 | ||
| AB109092.1 | BAC81437.1 | |||
| AK041947.1 | ||||
| BC111906.1 | AAI11907.1 |
More Information
Set Membership: |
|

Genomic Map