Assay Details
Target Gene Details
Entrez Gene ID: | 81601 |
Gene Name: | K(lysine) acetyltransferase 5 |
Gene Aliases: |
AI839539, CPLA2, Htatip, Htatip1, PLIP, Tip55, Tip60 |
Location: |
Chr.19:5603014-5610232 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kat5 | NM_001199247.1 | 9 | 912 | NP_001186176.1 |
| NM_001199248.1 | 8 | 1011 | NP_001186177.1 | |
| NM_001199249.1 | 8 | 1011 | NP_001186178.1 | |
| NM_178637.2 | 10 | 1068 | NP_848752.1 | |
| NR_037603.1 | 7 | 944 | ||
| AB055409.1 | 7 | 922 | BAC53807.1 | |
| AF528194.1 | 6 | 622 | AAN77140.1 | |
| AF528195.1 | 9 | 1003 | AAN77141.1 | |
| AF528196.1 | 8 | 847 | AAN77142.1 | |
| AK143674.1 | 8 | 963 | ||
| AK146352.1 | 10 | 1025 | BAE27104.1 | |
| AK159568.1 | 8 | 944 | ||
| AK160077.1 | 8 | 944 | ||
| AK205910.1 | ||||
| AK219598.1 | ||||
| BC110675.1 | 10 | 1009 | AAI10676.1 | |
| BC129967.1 | 8 | 846 | ||
| BC129968.1 | 8 | 847 | ||
| CA317697.1 | 3 | 306 | ||
| DQ139980.1 | 9 | 1003 | AAZ67923.1 |
More Information
Set Membership: |
|

Genomic Map