Assay Details
Target Gene Details
Entrez Gene ID: | 81601 |
Gene Name: | K(lysine) acetyltransferase 5 |
Gene Aliases: |
AI839539, CPLA2, Htatip, Htatip1, PLIP, Tip55, Tip60 |
Location: |
Chr.19:5603014-5610232 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kat5 | NM_001199247.1 | NP_001186176.1 | ||
| NM_001199248.1 | NP_001186177.1 | |||
| NM_001199249.1 | NP_001186178.1 | |||
| NM_178637.2 | NP_848752.1 | |||
| NR_037603.1 | ||||
| AB055409.1 | BAC53807.1 | |||
| AF528194.1 | AAN77140.1 | |||
| AF528195.1 | AAN77141.1 | |||
| AF528196.1 | AAN77142.1 | |||
| AK143674.1 | ||||
| AK146352.1 | BAE27104.1 | |||
| AK159568.1 | ||||
| AK160077.1 | ||||
| AK206382.1 | ||||
| AK219943.1 | ||||
| BC110675.1 | AAI10676.1 | |||
| BC129967.1 | ||||
| BC129968.1 | ||||
| DQ139980.1 | AAZ67923.1 |
More Information
Set Membership: |
|

Genomic Map