Assay Details
Target Gene Details
Entrez Gene ID: | 18399 |
Gene Name: | solute carrier family 22 (organic anion transporter), member 6 |
Gene Aliases: |
NKT, Oat1, Orctl1, mOat1 |
Location: |
Chr.19:8617996-8628299 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc22a6 | NM_008766.3 | 3 | 827 | NP_032792.2 |
| AK035971.1 | 3 | 645 | BAC29261.1 | |
| BC021647.1 | 3 | 795 | AAH21647.1 | |
| U52842.1 | 3 | 708 | AAC53112.1 |
More Information
Set Membership: |
|

Genomic Map