Assay Details
Target Gene Details
Entrez Gene ID: | 67717 |
Gene Name: | lipase, gastric |
Gene Aliases: |
2310051B21Rik, AV082900 |
Location: |
Chr.19:33919441-33976813 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
More Information
Set Membership: |
|

Genomic Map