Assay Details
Target Gene Details
Entrez Gene ID: | 58205 |
Gene Name: | programmed cell death 1 ligand 2 |
Gene Aliases: |
B7-DC, Btdc, F730015O22Rik, PD-L2 |
Location: |
Chr.19:29406240-29472927 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pdcd1lg2 | NM_021396.2 | NP_067371.1 | ||
| AF142780.2 | AAD33892.1 | |||
| AK089369.1 | BAC40858.1 | |||
| AK141111.1 | BAE24564.1 | |||
| AK155002.1 | BAE32986.1 | |||
| BC104138.1 | AAI04139.1 | |||
| BC104139.1 | AAI04140.1 | |||
| BY749628.1 |
More Information
Set Membership: |
|

Genomic Map