Assay Details
Target Gene Details
Entrez Gene ID: | 18150 |
Gene Name: | nucleoplasmin 3 |
Gene Aliases: |
Nub1 |
Location: |
Chr.19:45747734-45749563 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Npm3 | NM_008723.1 | NP_032749.1 | ||
| AK011749.1 | BAB27818.1 | |||
| AK146603.1 | BAE27297.1 | |||
| AK157198.1 | BAE33994.1 | |||
| AK171848.1 | BAE42695.1 | |||
| BC048491.1 | AAH48491.1 | |||
| U64450.1 | AAC53205.1 |
More Information
Set Membership: |
|

Genomic Map