Assay Details
Target Gene Details
Entrez Gene ID: | 58988 |
Gene Name: | ribosomal protein S6 kinase, polypeptide 2 |
Gene Aliases: |
70kDa, S6K2 |
Location: |
Chr.19:4156792-4163383 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rps6kb2 | NM_021485.2 | NP_067460.1 | ||
| AJ007938.1 | CAA07774.1 | |||
| AK014412.1 | BAB29335.1 | |||
| AK156361.1 | ||||
| BC005694.1 | AAH05694.1 | |||
| BC051631.1 | AAH51631.1 | |||
| BC066842.1 | AAH66842.1 |
More Information
Set Membership: |
|

Genomic Map