Assay Details
Target Gene Details
Entrez Gene ID: | 98170 |
Gene Name: | transmembrane protein 132A |
Gene Aliases: |
6720481D13Rik, Hspa5bp1, Orai1, R74613 |
Location: |
Chr.19:10857829-10869935 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem132a | NM_133804.2 | NP_598565.2 | ||
| AK032956.1 | ||||
| AK036651.1 | ||||
| AK048556.1 | BAC33373.1 | |||
| AK164646.1 | ||||
| AK173207.1 | BAD32485.1 | |||
| BC006896.1 |

Genomic Map