Assay Details
Target Gene Details
Entrez Gene ID: | 104401 |
Gene Name: | pecanex-like 3 (Drosophila) |
Gene Aliases: |
mKIAA4073 |
Location: |
Chr.19:5664635-5688908 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pcnxl3 | NM_144868.3 | 14 | 3427 | NP_659117.2 |
| AF237953.1 | 12 | 1444 | AAL62024.1 |

Genomic Map