Assay Details
Target Gene Details
Entrez Gene ID: | 67155 |
Gene Name: | SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 2 |
Gene Aliases: |
2610209L14Rik, SNF2alpha, Snf2l2, brahma, brm |
Location: |
Chr.19:26605089-26778321 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Smarca2 | NM_011416.2 | 6 | 1249 | NP_035546.2 |
| AK020092.1 | 6 | 1214 | BAB31990.1 | |
| AK147322.1 | 6 | 1450 | BAE27845.1 | |
| BC075641.1 | 6 | 1231 | AAH75641.1 |
More Information
Set Membership: |
|

Genomic Map