Assay Details
Target Gene Details
Entrez Gene ID: | 70942 |
Gene Name: | RIKEN cDNA 4931403E22 gene |
Gene Aliases: |
- |
Location: |
Chr.19:26768801-26823907 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4931403E22Rik | NR_045306.1 | |||
| AK016426.1 | ||||
| BY716074.1 |
Target Gene Details
Entrez Gene ID: | 67155 |
Gene Name: | SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 2 |
Gene Aliases: |
2610209L14Rik, SNF2alpha, Snf2l2, brahma, brm |
Location: |
Chr.19:26605089-26778321 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 32 - Exon 33 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Smarca2 | NM_011416.2 | NP_035546.2 | ||
| NM_026003.2 | NP_080279.1 | |||
| AK009309.1 | ||||
| AK011935.1 | BAB27925.1 | |||
| AK147322.1 | BAE27845.1 | |||
| BC004653.1 | AAH04653.1 | |||
| BC022975.1 | AAH22975.1 | |||
| BC056199.1 | AAH56199.1 | |||
| BC075641.1 | AAH75641.1 |
More Information
Set Membership: |
|

Genomic Map