Assay Details
Target Gene Details
Entrez Gene ID: | 26412 |
Gene Name: | mitogen-activated protein kinase kinase kinase kinase 2 |
Gene Aliases: |
AI385662, BL44, GCK, Rab8ip |
Location: |
Chr.19:6341116-6355619 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 21 - Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Map4k2 | NM_001291787.1 | NP_001278716.1 | ||
| NM_009006.3 | NP_033032.1 | |||
| NR_117093.1 | ||||
| AK011708.1 | ||||
| AK042262.1 | ||||
| AK051036.1 | BAC34507.1 | |||
| AK078526.1 | ||||
| AK154164.1 | ||||
| AK154323.1 | BAE32514.1 | |||
| AK216809.1 | ||||
| BC116286.1 | ||||
| BC116287.1 | ||||
| U50595.1 | AAC52571.1 |
Target Gene Details
Entrez Gene ID: | 102465598 |
Gene Name: | microRNA 6989 |
Gene Aliases: |
Gm27885, mmu-mir-6989 |
Location: |
Chr.19:6346480-6346544 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mir6989 | NR_105955.1 |

Genomic Map