Assay Details
Target Gene Details
Entrez Gene ID: | 18938 |
Gene Name: | protein phosphatase 1, regulatory (inhibitor) subunit 14B |
Gene Aliases: |
AOM172, PHI-1, PLCB3N, Png |
Location: |
Chr.19:6975048-6977324 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppp1r14b | NM_008889.2 | NP_032915.2 | ||
| AK141152.1 | ||||
| AK165792.1 | BAE38382.1 | |||
| BC082545.1 | AAH82545.2 | |||
| BC125650.1 | ||||
| BC125654.1 | ||||
| CF739461.1 | ||||
| X97490.1 | CAA66121.1 |
More Information
Set Membership: |
|

Genomic Map