Assay Details
Target Gene Details
Entrez Gene ID: | 225887 |
Gene Name: | NADH dehydrogenase (ubiquinone) Fe-S protein 8 |
Gene Aliases: |
BC021616, CI-23kD, TYKY |
Location: |
Chr.19:3908863-3912774 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ndufs8 | NM_001271443.1 | NP_001258372.1 | ||
| NM_001271444.1 | NP_001258373.1 | |||
| NM_144870.5 | NP_659119.2 | |||
| AK135069.1 | BAE22408.1 | |||
| AK149674.1 | BAE29018.1 | |||
| AK153182.1 | BAE31784.1 | |||
| AK153900.1 | BAE32243.1 | |||
| AK164901.1 | BAE37960.1 | |||
| AK167907.1 | BAE39914.1 | |||
| BC021616.1 | AAH21616.2 | |||
| BC086766.1 | AAH86766.1 |
More Information
Set Membership: |
|

Genomic Map