Assay Details
Target Gene Details
Entrez Gene ID: | 59028 |
Gene Name: | RNA terminal phosphate cyclase-like 1 |
Gene Aliases: |
2310040A02Rik, AI789745, C76567, RPCL1, Rnac |
Location: |
Chr.19:29101375-29143851 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rcl1 | NM_021525.2 | NP_067500.1 | ||
| AJ276895.1 | CAB89817.1 | |||
| AK009709.1 | BAB26454.1 | |||
| AK012585.1 | ||||
| AK077751.1 | ||||
| AK153377.1 | ||||
| AK168153.1 | BAE40118.1 | |||
| BC004574.1 | AAH04574.1 |
More Information
Set Membership: |
|

Genomic Map