Assay Details
Target Gene Details
Entrez Gene ID: | 68323 |
Gene Name: | nudix (nucleoside diphosphate linked moiety X)-type motif 22 |
Gene Aliases: |
0610006K04Rik, AI851793, AW545547 |
Location: |
Chr.19:6993019-6996115 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nudt22 | NM_026675.2 | NP_080951.1 | ||
| AK002252.1 | BAB21966.1 | |||
| BC019768.1 | AAH19768.1 |
More Information
Set Membership: |
|

Genomic Map