Assay Details
Target Gene Details
Entrez Gene ID: | 320376 |
Gene Name: | BCL6 co-repressor-like 1 |
Gene Aliases: |
6720425J07Rik, B930011H20Rik |
Location: |
Chr.X:48341358-48408571 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bcorl1 | NM_178782.4 | NP_848897.3 | ||
| AK031119.1 | BAC27262.1 | |||
| AK054259.1 | BAC35708.1 | |||
| AK081004.1 | BAC38112.1 |

Genomic Map