Assay Details
Target Gene Details
Entrez Gene ID: | 74405 |
Gene Name: | EF-hand domain (C-terminal) containing 2 |
Gene Aliases: |
4933407D04Rik, mRib72-2 |
Location: |
Chr.X:17132049-17320461 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Efhc2 | NM_028916.4 | NP_083192.2 | ||
| AK016714.1 | BAB30393.1 | |||
| AK029501.1 | BAC26479.1 | |||
| BC120659.1 | ||||
| BC125625.1 |
More Information
Set Membership: |
|

Genomic Map