Assay Details
Target Gene Details
Entrez Gene ID: | 56292 |
Gene Name: | N(alpha)-acetyltransferase 10, NatA catalytic subunit |
Gene Aliases: |
2310039H09Rik, Ard1, Ard1a, Te2 |
Location: |
Chr.X:73916870-73921944 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Naa10 | NM_001177965.1 | NP_001171436.1 | ||
| NM_019870.3 | NP_063923.1 | |||
| AJ627039.1 | CAF25308.1 | |||
| AK003806.1 | BAE43151.1 | |||
| AK009697.1 | BAC25266.1 | |||
| AK014469.1 | BAB29373.1 | |||
| AK078700.1 | BAC37364.1 | |||
| AY183136.1 | AAO66339.1 | |||
| AY183137.1 | ||||
| BC027219.1 | AAH27219.1 | |||
| BY101790.1 |
More Information
Set Membership: |
|

Genomic Map