Assay Details
Target Gene Details
Entrez Gene ID: | 171486 |
Gene Name: | CD99 antigen-like 2 |
Gene Aliases: |
AW548191, Mic2l1, Xap89 |
Location: |
Chr.X:71420060-71492869 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cd99l2 | NM_001199349.1 | NP_001186278.1 | ||
| NM_138309.3 | NP_612182.1 | |||
| AK031264.1 | ||||
| AK080225.1 | BAC37853.1 | |||
| AK166275.1 | BAE38674.1 | |||
| AY078163.1 | AAL86615.1 | |||
| BC031736.1 | AAH31736.1 | |||
| BY727540.1 | ||||
| EF516990.1 |
More Information
Set Membership: |
|

Genomic Map