Assay Details
Target Gene Details
Entrez Gene ID: | 245536 |
Gene Name: | predicted gene 614 |
Gene Aliases: |
Il2rg |
Location: |
Chr.X:101261377-101263992 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm614 | NM_001033362.2 | NP_001028534.1 | ||
| AK131750.1 | BAE20796.1 | |||
| BC147523.1 | ||||
| BC147537.1 |
More Information
Set Membership: |
|

Genomic Map