Assay Details
Target Gene Details
Entrez Gene ID: | 67391 |
Gene Name: | FUN14 domain containing 2 |
Gene Aliases: |
4833415N24Rik, DC44, HCBP6 |
Location: |
Chr.X:75382399-75397158 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fundc2 | NM_026126.4 | NP_080402.3 | ||
| AK013460.1 | BAB28867.1 | |||
| AK014705.1 | BAB29513.1 | |||
| AK031214.1 | BAC27304.1 | |||
| AY234858.1 | AAO89275.1 | |||
| BC042794.1 | AAH42794.1 | |||
| BC049566.1 | AAH49566.2 | |||
| BC061464.1 | AAH61464.1 | |||
| BY124891.1 |

Genomic Map